Learn More
Kingfisher Biotech Inc Rabbit CXCL1 ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891792
Encompass_Preferred
Rabbit CXCL1 ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1792 Supplier Kingfisher Biotech Inc Supplier No. SPOT2505U004
Rabbit CXCL1 ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA