Learn More
Raybiotech Inc G-Series Rabbit Cytokine Array 1 Kit.
Supplier:  Raybiotech Inc GSLCYT11
Encompass_Preferred
Semi-quantitative measurement of 10 rabbit cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-8 (CXCL8), IL-17A, IL-21 (IL-17C), Leptin, MIP-1 beta (CCL4), MMP-9, NCAM-1 (CD56), TNF alpha
Catalog No. 50-195-0434