Learn More
Raybiotech Inc G-Series Rabbit Cytokine Array 2 Kit.
Supplier:  Raybiotech Inc GSLCYT21
Encompass_Preferred
Detects 10 Rabbit Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: AMSH, BLC (CXCL13), FLRG, IL-13, IL-17F, IL-1RA, RANTES (CCL5), SCF, TWEAK R, VEGF-A (VEGF)
Catalog No. 50-195-0436