Learn More
Raybiotech Inc G-Series Rabbit Cytokine Array 3 Kit.
Catalog No. 501950438
Encompass_Preferred
Detects 10 Rabbit Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 6Ckine (CCL21), Angiopoietin-1 (ANG-1), ANGPTL3, BLMH, CMASP3, Contactin-1, Matrilin-3, Notch-3, SREC-II (SCARF2), Thrombospondin-2 (TSP-2)
Catalog No. 50-195-0438 Supplier Raybiotech Inc Supplier No. GSLCYT31
Detects 10 Rabbit Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 6Ckine (CCL21), Angiopoietin-1 (ANG-1), ANGPTL3, BLMH, CMASP3, Contactin-1, Matrilin-3, Notch-3, SREC-II (SCARF2), Thrombospondin-2 (TSP-2)