Learn More
Raybiotech Inc Quantibody Rabbit Cytokine Antibody Array 30 Kit.
Supplier:  Raybiotech Inc QALCAA302
Encompass_Preferred
A combination of 3 non-overlapping arrays to quantitatively measure 30 Rabbit cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: 6Ckine, AMSH, Angiopoietin-1, ANGPTL3, BLC, BLMH, Complement MASP3, Contactin-1, FLRG, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-13, IL-17A, IL-17F, IL-21, IL-8, Leptin, Matrilin-3, MIP-1 beta, MMP-9, NCAM-1, Notch-3, RANTES, SCF, SREC-II, Thrombospondin-2, TNF alpha, TWEAK R, VEGF-A
Catalog No. 50-195-0860