Learn More
Biomatik Corporation Rabbit E-selectin (E-selectin) ELISA Kit
Supplier:  Biomatik Corporation EKU07246
Encompass_Preferred
Rabbit E-selectin (E-selectin) ELISA Kit for detecting E-selectin in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27113. Gene ID: 100009151. Target Synonyms: CD62E; CD62-E; E-Selectin; ELAM; ELAM1; ESEL; SEL-E; E-LAM; E-LAM1; E-SEL; LECAM2; Endothelial Leukocyte Adhesion Molecule 1; CD62 Antigen-Like Family Member E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6154