Learn More
Sino Biological Recombinant Rabbit CD64A Protein (His Tag)
Supplier:  Sino Biological 65010T08H1
Encompass_Preferred
A DNA sequence encoding the rabbit FCGR1A (XP_008262732.1) (Met1-Phe291) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2928