Learn More
Cusabio Technology LLC RABBIT FGF2 ELISA KIT
Catalog No. 5000050494
Encompass_Preferred
This kit detects FGF2 (basic fibroblast growth factor) in serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 7 8 pg/mL to 500 pg/mL with a sensitivity of 1 95 pg/mL enabling precise measurement of low-abundance samples The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 L per test Absorbance is measured at 450 nm ensuring reliable and reproducible results for both research and diagnostic applications Designed for flexibility the kit is suitable for analyzing diverse biological matrices while maintaining high specificity and accuracy in quantifying FGF2 levels
Catalog No. 50-000-50494 Supplier Cusabio Technology LLC Supplier No. CSBE06938RB
This kit detects FGF2 (basic fibroblast growth factor) in serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 7 8 pg/mL to 500 pg/mL with a sensitivity of 1 95 pg/mL enabling precise measurement of low-abundance samples The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 L per test Absorbance is measured at 450 nm ensuring reliable and reproducible results for both research and diagnostic applications Designed for flexibility the kit is suitable for analyzing diverse biological matrices while maintaining high specificity and accuracy in quantifying FGF2 levels