Learn More
Innovative Research Inc Rabbit Fibrinogen ELISA Kit
Supplier:  Innovative Research Inc IRBFBGKT
Encompass_Preferred
This rabbit fibrinogen antigen assay is intended for the quantitative determination of total fibrinogen antigen in rabbit plasma and serum. For research use only. Strip well format. Reagents for up to 96 tests.
Catalog No. 50-203-6135