Learn More
American Research Products Inc Rabbit Fibronectin FN1 ELISA
Catalog No. 5000327696
Encompass
Rabbit Fibronectin (FN1) ELISA kit from CUSABIO Detection Range 1 25 ng/mL-20 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27696 Supplier American Research Products Inc Supplier No. CSBEL008759RB
Rabbit Fibronectin (FN1) ELISA kit from CUSABIO Detection Range 1 25 ng/mL-20 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma tissue homogenates Method Competitive