Learn More
ACROBiosystems Rabbit FMC63 960tests
Supplier:  ACROBiosystems RABP001960TESTS
Encompass_Preferred
FMC63 is an IgG2a mouse monoclonal antibody specific for CD19 which is a target for the immunotherapy of B lineage leukaemias and lymphomas. FMC63 scFv is the most commonly used ectodomain component of CD19-specific CARs. So far most of reported CART19 trials contain the anti-CD19 scFv derived from FMC63 including the two FDA-approved CARs Kymriah and Yescarta.Anti-fmc63 scFv antibody can specifically bind to the antigen recognition epitope of fmc63 scFv on anti-CD19 car and it shows high specificity and sensitivityits used to detect the expression of fmc63 scFv derived car.
Catalog No. 50-253-4270
Add to Cart
Request a Quote