Learn More
ACROBiosystems Anti-CD19 (FMC63) CAR Immunogenicity ELISA Kit, Bridging-ELISA, 96 Tests
Catalog No. 502331316
Encompass_Preferred
The Anti-CD19 (FMC63) CAR Immunogenicity ELISA Kit utilizes a bridging-ELISA methodology to detect and quantitatively determine anti-CD19 (FMC63) antibodies in CAR immunogenicity assays. This kit provides a reliable solution for monitoring anti-drug antibody (ADA) levels, which is essential for evaluating potential immune responses in CAR T-cell therapy.
  • Designed for sensitive detection using the bridging-ELISA method.
  • Sensitivity of 19 ng/mL.
  • Includes high-bind plate, mouse FMC63 scFv, standard, biotinylated mouse FMC63 scFv, and streptavidin-HRP.
  • Provided with necessary reagents including coating, blocking, and washing buffers, substrate solution, and stop solution.
  • Supports quality management under ISO and GMP standards.
Catalog No. 50-233-1316 Supplier ACROBiosystems Supplier No. RABP00196TESTS
The Anti-CD19 (FMC63) CAR Immunogenicity ELISA Kit utilizes a bridging-ELISA methodology to detect and quantitatively determine anti-CD19 (FMC63) antibodies in CAR immunogenicity assays. This kit provides a reliable solution for monitoring anti-drug antibody (ADA) levels, which is essential for evaluating potential immune responses in CAR T-cell therapy.
  • Designed for sensitive detection using the bridging-ELISA method.
  • Sensitivity of 19 ng/mL.
  • Includes high-bind plate, mouse FMC63 scFv, standard, biotinylated mouse FMC63 scFv, and streptavidin-HRP.
  • Provided with necessary reagents including coating, blocking, and washing buffers, substrate solution, and stop solution.
  • Supports quality management under ISO and GMP standards.