Learn More
American Research Products Inc Rabbit follicle stimulating ho
Catalog No. 5000326284
Encompass
Rabbit follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26284 Supplier American Research Products Inc Supplier No. CSBE06918RB
Rabbit follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma tissue homogenates Method Sandwich