Learn More
Biomatik Corporation Rabbit Glutamate Receptor, Ionotropic, AMPA 1 (GRIA1) ELISA Kit
Supplier:  Biomatik Corporation EKU10867
Encompass_Preferred
Rabbit Glutamate Receptor, Ionotropic, AMPA 1 (GRIA1) ELISA Kit for detecting GRIA1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A5F9D438. Target Synonyms: GLUH1; GLUR1; GLURA; HBGR1; GluR-K1; AMPA-selective glutamate receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6167