Learn More
American Research Products Inc Rabbit Growth Hormone GH ELI
Catalog No. 5000329562
Encompass
Rabbit Growth Hormone (GH) ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-100 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29562 Supplier American Research Products Inc Supplier No. CSBE15021RB
Rabbit Growth Hormone (GH) ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-100 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Competitive