Learn More
Euthanex Rabbit/Guinea Pig Breather Mask
Catalog No. NC1880352
Encompass_Preferred
Versaflex Breather Mask efficiently delivers anesthetic gas to the animal during procedure. The breather is ideal for use with rabbits, ferrets, and guinea pigs. Input tube delivers anesthetizing gas to the animal and an exhaust tube directs the flow of unused gas to a charcoal filter. A flexible membrane seal eliminates gas leakage and protects personnel from gas exposure.
Catalog No. NC1880352 Supplier Euthanex Supplier No. EZ105A
Versaflex Breather Mask efficiently delivers anesthetic gas to the animal during procedure. The breather is ideal for use with rabbits, ferrets, and guinea pigs. Input tube delivers anesthetizing gas to the animal and an exhaust tube directs the flow of unused gas to a charcoal filter. A flexible membrane seal eliminates gas leakage and protects personnel from gas exposure.