Learn More
World Precision Instrument Rabbit/Guinea Pig Nosecone for Anesthesia
SDP
Catalog No. 502538848
Encompass
EZ Anesthesia nosecones are custom-designed for small animals, ensuring a tight fit and minimal dead space. They are available in three sizes to fit mice, rats and rabbits. Nosecones include rubber plugs to close off gas supply when not in use. Nosecone plugs are also available separately (EZ-119).
Catalog No. 50-253-8848 Supplier World Precision Instrument Supplier No. EZ115
EZ Anesthesia nosecones are custom-designed for small animals, ensuring a tight fit and minimal dead space. They are available in three sizes to fit mice, rats and rabbits. Nosecones include rubber plugs to close off gas supply when not in use. Nosecone plugs are also available separately (EZ-119).