Learn More
Euthanex Rabbit/ Guinea Pig Nosecone
Catalog No. NC1880354
Encompass_Preferred
The Rabbit/Guinea Pig Nosecone is custom-designed for small animals, ensuring a tight fit and minimal dead space. This is a component part of the E-Z Anesthesia breathing devices. Nosecone includes rubber plug to close off gas supply when not in use.
Catalog No. NC1880354 Supplier Euthanex Supplier No. EZ115
The Rabbit/Guinea Pig Nosecone is custom-designed for small animals, ensuring a tight fit and minimal dead space. This is a component part of the E-Z Anesthesia breathing devices. Nosecone includes rubber plug to close off gas supply when not in use.