Learn More
Electron Microscopy Sciences Rabbit Heart Matrix Adult, 2 mm Slices
SDP
Catalog No. 5028369
Encompass_Preferred
Constructed from high grade zinc to allow for rapid temperature equilibration, easy cleaning, and sterilization.
Size: 2 mm Slices
Catalog No. 50-283-69 Supplier Electron Microscopy Sciences Supplier No. 69051C
Constructed from high grade zinc to allow for rapid temperature equilibration, easy cleaning, and sterilization.
Size: 2 mm Slices