Learn More
Biomatik Corporation Rabbit Heat Shock Protein 27 (Hsp27) ELISA Kit
Supplier:  Biomatik Corporation EKU11678
Encompass_Preferred
Rabbit Heat Shock Protein 27 (Hsp27) ELISA Kit for detecting Hsp27 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1T3V2. Gene ID: 100354523. Target Synonyms: HSPB1; HSP-B1; CMT2F; HSP28; Hsp25; Heat Shock 27kDa Protein 1; 28 kDa heat shock protein; Heat shock protein beta-1; Estrogen-regulated 24 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8100