Learn More
Abcam Rabbit specific HRP/DAB Detection IHC Kit
Catalog No. NC3780275
Encompass_Preferred
Rabbit-specific HRP/DAB IHC detection is a complete kit to detect a primary antibody raised in rabbit, using HRP-labeled-streptavidin and a biotinylated anti-rabbit secondary antibody. - Cited in over 200 publications - Includes peroxidase and protein blocking reagents - Produces clear brown staining with DAB - Ensures strong and specific antigen detection
Catalog No. NC3780275 Supplier Abcam Supplier No. AB64261
Rabbit-specific HRP/DAB IHC detection is a complete kit to detect a primary antibody raised in rabbit, using HRP-labeled-streptavidin and a biotinylated anti-rabbit secondary antibody. - Cited in over 200 publications - Includes peroxidase and protein blocking reagents - Produces clear brown staining with DAB - Ensures strong and specific antigen detection