Learn More
Kingfisher Biotech Inc Rabbit IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0136U005
Encompass
The Rabbit IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. The Rabbit IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit IFN gamma Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: QDTLTRETEH LKAYLKANTS DVANGGPLFL NILRNWKEES DNKIIQSQIV SFYFKLFDNL KDHEVIKKSM ESIKEDIFVK FFNSNLTKMD DFQNLTRISV DDRLVQRKAV SELSNVLNFL SPKSNLKKRK RSQTLFRGRR ASKY (144)) (Gene ID: 100008602). For research use only. Made in the USA
Catalog No. 50-241-3635