Learn More
Vector Laboratories Rabbit IgG (Control Antibody) 5 mg
SDP
Supplier:  Vector Laboratories I10005
Encompass
This non-specific rabbit IgG preparation is intended for use as a negative control for primary antibodies made in rabbit. The antibody has been purified from pooled serum of healthy adult animals and contains a spectrum of the IgG subclasses. The control antibody should be applied to the tissue section at the same concentration as the primary antibody to indicate whether staining is specific for the antigen or is nonspecific adsorption of primary antibody to tissue sites. Supplied as a lyophilized powder.
Catalog No. NC9593121