Learn More
Innovative Research Inc Rabbit IgG Affinity Purified Low Endotoxin Lyophilized 50mg
Supplier:  Innovative Research Inc IRBIGGAPLXLY50MG
Encompass_Preferred
Rabbit IgG Affinity Purified Low Endotoxin from Innovative Research has been purified from normal Rabbit Serum using Protein A affinity chromatography then endotoxin depleted and sterile filtered. This is a lyophilized powder buffered in 0.015M K Phosphate; 0.027M Na Phosphate; 0.15M NaCl; pH 7.2, with a purity of >95%.
Catalog No. 50-203-6543