Learn More
Genscript Corporation Rabbit IgG Control (Whole Molecule), Purified
Catalog No. 502271945
Encompass
GenScript Purified Rabbit IgG (Whole Molecule) Control is highly purified from normal rabbit serum by Protein G chromatography.GenScript Purified Rabbit IgG (Whole Molecule) Control is suitable for use as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, immunobloting (Dot blot and Western blot), immunoprecipitation, immunodiffusion, and immunoelectrophoresis.It also may be used as antigen or ligand in immunochemical reactions and conjugations.
This item is not returnable. View return policy
Catalog No. 50-227-1945 Supplier Genscript Corporation Supplier No. A01008
GenScript Purified Rabbit IgG (Whole Molecule) Control is highly purified from normal rabbit serum by Protein G chromatography.GenScript Purified Rabbit IgG (Whole Molecule) Control is suitable for use as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, immunobloting (Dot blot and Western blot), immunoprecipitation, immunodiffusion, and immunoelectrophoresis.It also may be used as antigen or ligand in immunochemical reactions and conjugations.