Learn More
Rockland Immunochemicals Rabbit IgG (H&L) Antibody Dylight™ 680 Conjugated 500 µg
SDP
Supplier:  Rockland Immunochemicals 6111440020.5
Encompass
This product is designed for immunofluorescence microscopy fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis including multicolor imaging utilizing various commercial platforms.
Catalog No. 50-194-3921