Learn More
Abcam Rabbit IgG ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1874001X96TESTS
Encompass_Preferred
Rabbit IgG ELISA Kit is a single-wash 90-min Simplestep used to quantify Rabbit IgG with a sensitivity of 0.23 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1932461