Learn More
Rockland Immunochemicals Rabbit IgG F(c) Antibody Peroxidase Conjugated Pre-Adsorbed 2 mg
SDP
Supplier:  Rockland Immunochemicals 6111323
Encompass
Anti-Rabbit IgG F(c) peroxidase antibody generated in goat is a proteolytic fragment of immunoglobulin G (IgG) obtained by limited digestion with the enzyme papain under controlled conditions of temperature time and pH. Receptors bind the Fc portion of rabbit IgG and often this fragment is removed from immunoglobulins to minimize receptor binding and lower background reactivity.
Catalog No. 50-194-3920