Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Franction to Hamster IgG, Secondary Antibody, MP Biomedicals™

Catalog No. MP0855398 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0855398 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0855398 Supplier MP Biomedicals, Inc Supplier No. 0855398
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit anti- Hamster IgG (Whole Molecule) Secondary Antibody

  • Product is the lyophilized powder of rabbit IgG fraction to hamster IgG (whole molecule) and buffer salts.
  • The IgG fraction is prepared from the specific rabbit antiserum by delipidation, ammonium sulfate fractionation and anion exchange chromatography
  • This product is suitable for use as a precipitating antibody. It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques.
  • This product is suitable for conjugation with enzymes, radiolabels, fluorochromes or other markers, and for attachment to solid supports for use as an immunoadsorbant.
  • This product is suitable for use as a primary reagent in enzyme immunoassays, fluorescein immunoassays and blot, cell or tissue immunostaining.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry, ELISA
Conjugate Unconjugated
Description Product is the lyophilized powder of rabbit IgG fraction to hamster IgG (whole molecule) and buffer salts
Host Species Rabbit
Purification Method ammonium sulfate fractionation and anion exchange chromatography
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute product with 5.0mL of deionized or distilled water.gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material.
Target Species Hamster
Content And Storage Store lyophilized product at 2-8°C. For long term storage, aliquot and store reconstituted product frozen at -20°C or lower.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.