Learn More
Rockland Immunochemicals Rabbit IgG (H&L) Antibody Biotin Conjugated 2 mg
SDP
Supplier:  Rockland Immunochemicals 6111602
Encompass
Anti-Rabbit IgG (H&L) Biotin Antibody generated in goat detects reactivity to Rabbit IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3925