Learn More
Rockland Immunochemicals Rabbit IgG (H&L) Antibody CY5 Conjugated Pre-adsorbed 1 mg
SDP
Supplier:  Rockland Immunochemicals 611110122
Encompass
Anti-Rabbit IgG Antibody CY5 generated in goat detects rabbit IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. This Anti-Rabbit IgG (H&L) is conjugated to CY5.
Catalog No. 50-194-3915