Learn More
Sigma Aldrich Fine Chemicals Biosciences Rabbit IgG HRP Linked fab2
Catalog No. 502904232
Encompass
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 C to 8 C.
Catalog No. 50-290-4232 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GENA93401ML
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 C to 8 C.