Learn More
Sigma Aldrich Fine Chemicals Biosciences Rabbit IgG HRP Linked Whol
Catalog No. 502901090
Encompass
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 C to 8 C.
Catalog No. 50-290-1090 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GENA934100UL
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 C to 8 C.