Learn More
Enzo Life Sciences Goat anti-rabbit IgG, polyclonal antibody (HRP conjugate) (1 ml)
SDP
Supplier:  Enzo Life Sciences ADISAB300J
Encompass_Preferred
Host: Goat. Immunogen: Purified rabbit IgG (whole molecule). Specificity: Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule rabbit IgG. It also reacts with the light chains of other rabbit immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Species Reactivity: Rabbit. Applications: ELISA, IHC, WB. Recommended dilutions: ELISA (1:2,500-50,000). Immunohistochemistry (1:250-2500, Western Blot (1:5,000-100,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In PBS, pH 7.6, containing 7.5mg/mL BSA (IgG-free, protease-free) and 50% glycerol.
Catalog No. 50-667-388