Learn More
Raybiotech Inc Rabbit IL-1 beta (IL-1 F2) ELISA Kit
Supplier:  Raybiotech Inc ELLIL1B1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 45 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rabbit IL-1alpha, IL-8, IL-17A, IL-21, Leptin, MIP-1 beta, MMP-9, NCAM-1, TNF alpha
Catalog No. 50-194-9904