Learn More
Raybiotech Inc Rabbit IL-17A ELISA Kit
Supplier:  Raybiotech Inc ELLIL17A1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rabbit IL-17A
Catalog No. 50-194-9903