Learn More
Kingfisher Biotech Inc Rabbit IL-6 ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891793
Encompass_Preferred
Rabbit IL-6 ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1793 Supplier Kingfisher Biotech Inc Supplier No. SPOT2519U004
Rabbit IL-6 ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA