Learn More
Kingfisher Biotech Inc Swine TWEAK ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1432U003
Encompass_Preferred
The Swine TWEAK ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Swine TWEAK ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include TNFSF12, APO3L, DR3LG, TWEAK, TNLG4A, tumor necrosis factor superfamily member 12, TNF superfamily member 12. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5455