Learn More
Kingfisher Biotech Inc Bovine CTLA-4 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1432U004
Encompass_Preferred
The Bovine CTLA-4 ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Bovine CTLA-4 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Bovine CTLA-4 ELISA can also be used to measure Bison, Carabao, Wild Yak, Water Buffalo, and Zebu CTLA-4. Alternate names include CTLA4, ALPS5, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, IDDM12, cytotoxic T-lymphocyte associated protein 4. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5456