Learn More
Raybiotech Inc Rabbit IL-21 ELISA Kit
Supplier:  Raybiotech Inc ELLIL211
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 2.45 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rabbit IL-1alpha, IL-1beta, IL-8, IL-17A, Leptin, MIP-1beta, MMP-9, NCAM-1, TNF alpha
Catalog No. 50-194-9907