Learn More
Kingfisher Biotech Inc Rabbit IL-6 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1281U100
Encompass
The Rabbit IL-6 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit IL-6 applications are for cell culture, ELISA standard, and Western Blot Control. Rabbit IL-6 yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit IL-6 Specifications: (Molecular Weight: 24.5 kDa) (Amino Acid Sequence: FPTSAPVRED SNTKASPDKT LTPPGRTIES IRSILETIKE LRKEMCDHDV NCMNRKEALA EVNLHLPRLI EEDGCFPPAV NNETCLLRIT SGLMEFRMYL EHLQAKFRSD EENTRVSMVL KNIQHLIKTL RPKVKNLNEE ATLKPAVAVS LMENLQQKNQ WLKTTTIHFI LRGLTNFLEF TLRAVDLM (188)) (Gene ID: 100008733). For research use only. Made in the USA
Catalog No. 50-253-3445