Learn More
Kingfisher Biotech Inc Cynomolgus Monkey CXCL7 ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1392U003
Encompass_Preferred
The Cynomolgus Monkey CXCL7 ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Cynomolgus Monkey CXCL7 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Cynomolgus Monkey CXCL7 Do-It-Yourself ELISA can also be used to measure Macaque, Baboon, and Gelada CXCL7. Alternate names include PPBP, B-TG1, Beta-TG, CTAP-III, CTAP3, CTAPIII, CXCL7, LA-PF4, LDGF, MDGF, NAP-2, PBP, SCYB7, TC1, TC2, TGB, TGB1, THBGB, THBGB1, pro-platelet basic protein. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5443