Learn More
Kingfisher Biotech Inc Human CXCL7 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1392U004
Encompass_Preferred
The Human CXCL7 ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Human CXCL7 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Human CXCL7 ELISA can also be used to measure Chimpanzee CXCL7. Alternate names include PPBP, B-TG1, Beta-TG, CTAP-III, CTAP3, CTAPIII, CXCL7, LA-PF4, LDGF, MDGF, NAP-2, PBP, SCYB7, TC1, TC2, TGB, TGB1, THBGB, THBGB1, pro-platelet basic protein. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5444