Learn More
Sino Biological Rabbit IL18RAP Protein 100ug (Sino Biological)
Supplier:  Sino Biological 65170T08H100
Encompass_Preferred
A DNA sequence encoding the Rabbit IL18RAP (G1SQ92) (Met1-Val357) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2193