Learn More
American Research Products Inc Rabbit Immunoglobulin A IgA E
Catalog No. 5000326477
Encompass
Rabbit Immunoglobulin A IgA ELISA Kit from CUSABIO Detection Range 12 ng/mL-3000 ng/mL Sensitivity 3 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-26477 Supplier American Research Products Inc Supplier No. CSBE06946RB
Rabbit Immunoglobulin A IgA ELISA Kit from CUSABIO Detection Range 12 ng/mL-3000 ng/mL Sensitivity 3 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive