Learn More
Biomatik Corporation Rabbit Immunoglobulin E (IgE) ELISA Kit
Supplier:  Biomatik Corporation EKU10993
Encompass_Preferred
Rabbit Immunoglobulin E (IgE) ELISA Kit for detecting IgE in serum, plasma,, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 7.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: QGU21316.1. Target Synonyms: IGHE; Immunoglobulin Heavy Constant Epsilon; Ig epsilon chain C region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8253