Learn More
Bioworld Rabbit Immunoglobulin G Lyophilized Powder >= 97%, from US serum, 100 g
Supplier:  Bioworld 220701273
Encompass_Preferred
Specifications:Rabbit IgG, Lyophilized powderPurity: ≥98%Solubility: Reconstitute in distilled water or buffer of choice Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product, food additive or as a household chemical.
Catalog No. 50-253-989