Learn More
Crystal Chem Inc Rabbit Insulin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 90186
Encompass
The Rabit Insulin ELISA is used to quantify levels of rabbit insulin in plasma, serum, and cell culture using only a 5 µL sample volume. Highlights: Kits use only 5 µL sample. Dynamic range of 0.1- 64 ng/mL. High sensitivity of 50 pg/mL using 5µL sample.
Catalog No. 50-194-7930