Learn More
American Research Products Inc Rabbit Insulin like growth fac
Supplier:  American Research Products Inc CSBE06994RB
Encompass
Rabbit Insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26662