Learn More
Biomatik Corporation Rabbit Insulin, Ins Elisa Kit
Supplier:  Biomatik Corporation EKC31471
Encompass_Preferred
Rabbit Insulin, Ins Elisa Kit for detecting Insulin in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 8-100uIU/ml. Sensitivity: 5uIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01311; INS. Target Synonyms: INS; Insulin [Cleaved into: Insulin B chain; Insulin A chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9185